missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NoTrax™ Marble Sof-Tyle™ Anti-Fatigue Mat
Marble Sof-Tyle™ is an anti-fatigue mat with a heavy-duty vinyl stylish smooth marble-pattern design with a dense sponge backing that utilizes NoTrax™ exclusive UniFusion™ technology. Comes with RedStop™, a non-slip backing that eliminates slipping and sliding of the mat on smooth surfaces.
Supplier: NoTrax™ 470R0048BL
Description
- Marble Sof-Tyle™ is an anti-fatigue floor mat provides comfort and ergonomic support, while also complementing any deÌcor with a stylish smooth marble-pattern design
- The durable vinyl top surface is easy to sweep clean or damp mop and is combined with a dense closed-cell foam base that utilizes NoTrax™ exclusive UniFusion™ technology which virtually eliminates the possibility of de-lamination
- NoTrax™ laminate floor mats that come with RedStop™, a uniquely engineered non-slip backing technology, eliminate slipping and sliding of floor mats on smooth surface flooring such as tile, wood, marble, and treated concrete
- For dry applications such as lab testing area, check-out counters, order fulfillment lines and pharmacies
- The sloped beveled edges reduce tripping hazards and allow easy access on and off the mat
Specifications
| 75 x 4 ft. | |
| Black Smoke | |
| Placement in manufacturing plants, assembly lines, picking, packing and logistics warehouses, individual work stations | |
| 0.0127 m | |
| 1/2 in. | |
| Certified by the NFSI (National Floor Safety Institute) |
| 75 ft. | |
| 4 ft. | |
| Anti-fatigue Floor Mat | |
| Nitricell, PVC | |
| 1.2192 m | |
| 22.86 m |