missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline Hydrochloride 98.0+%, TCI America™

Catalog No. M25275G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25275G 5 g
M25271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25275G Supplier TCI America Supplier No. M25275G

Chemical Identifiers

CAS 10347-81-6
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.87
MDL Number MFCD00079464
InChI Key NZDMFGKECODQRY-UHFFFAOYSA-N
Synonym maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride
PubChem CID 71478
IUPAC Name hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride
SMILES [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Melting Point 242°C
Color White
Quantity 5 g
Formula Weight 313.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Maprotiline Hydrochloride
RTECSNumber KJ4555000
TSCA No