missing translation for 'onlineSavingsMsg'
Learn More

Maprotiline hydrochloride, 99%

Catalog No. AAJ6232106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6232106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6232106 Supplier Thermo Scientific Chemicals Supplier No. J6232106

Selective norepinephrine uptake inhibitor | CAS: 10347-81-6 | C20H24ClN | 313.87 g/mol

Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maprotiline Hydrochloride has also been shown to block apoptosis of spiny neurons and inhibit SLC6A2.

Solubility
Soluble to 50mg/ml in water. Soluble in ethanol at 10mM

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 10347-81-6
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.87
MDL Number MFCD00079464
InChI Key NZDMFGKECODQRY-UHFFFAOYSA-N
Synonym maprotiline hydrochloride, maprotiline hcl, ludiomil, psymion, maprotilline hcl, ciba 34276 ba, unii-7c8j54pvfi, deprilept, 9-gamma-methylaminopropyl-9,10-dihydro-9,10-ethanoanthracene hydrochloride
PubChem CID 71478
IUPAC Name hydrogen methyl(3-{tetracyclo[6.6.2.0²,⁷.0⁹,¹⁴]hexadeca-2,4,6,9,11,13-hexaen-1-yl}propyl)amine chloride
SMILES [H+].[Cl-].CNCCCC12CCC(C3=CC=CC=C13)C1=CC=CC=C21

Specifications

Melting Point 230°C to 232°C
Color White
Quantity 5 g
Beilstein 5463242
Merck Index 14,5748
Solubility Information Soluble to 50mg/ml in water; Soluble in ethanol at 10mM
Formula Weight 313.87
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Maprotiline hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 233-758-8
RTECSNumber KJ4555000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only