missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPRE2 Monoclonal Antibody (KT52), Invitrogen™
Rat Monoclonal Antibody
$1693.48 - $2847.38
Specifications
| Antigen | MAPRE2 |
|---|---|
| Clone | KT52 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
Description
MAPRE2 Monoclonal antibody specifically detects MAPRE2 in Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| MAPRE2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2 | |
| Recombinant mouse Mapre2 (proprietary range information). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MAPRE2 |
| KT52 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse, Rat, Monkey, Hamster | |
| Q15555, Q3B8Q0, Q8R001 | |
| 100765710, 10982, 212307, 679221 | |
| IgG2b | |
| Protein G | |
| Antibody |