missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Multispecies MAPK Family Activation InstantOne™ ELISA Kit

Catalog No. 501125032
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125032 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501125032 Supplier Invitrogen™ Supplier No. 858619511

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ELISA

The InstantOne™ ELISA is specifically engineered for accurate measurement of phosphorylated ERK 1/2 (human, mouse, hamster), p38 MAPK (human, mouse), and JNK 1/2 (human, mouse) in cell lysates. Detection of these phosphorylated molecules in rat is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.

MAP kinases play a significant role in many biological processes, including cell adhesion and spreading, cell differentiation and apoptosis. MAPKBP-1 (mitogen-activated protein kinase binding protein 1), also known as JNKBP-1, is a 1,514 amino acid protein that contains twelve WD repeats. Induced by TRAF2 (TNF receptor-associated factor 2) and Tak1 (TGF-β-activated kinase 1), MAPKBP-1 is thought to act an adaptor protein for NFκB (nuclear factor κ-B) activation. MAPKBP-1 interacts with JNK3 and may promote TRAF2 polyubiquitination. MAPKBP-1 exists as six alternatively spliced variants and is encoded by a gene located on human chromosome 15. Human chromosome 15 houses over 700 genes and comprises nearly 3% of the human genome. Angelman syndrome, Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are all associated with defects in chromosome 15-localized genes.

Specifications

Accession Number P27361, P28482, Q16539
Content And Storage • User Manual
• phospho-ERK 1/2 (Thr202/Tyr204, Thr182/Tyr187) Capture Antibody Reagent, 1 mL
• phospho-ERK 1/2 (Thr202/Tyr204, Thr182/Tyr187) Detection Antibody Reagent, 1 mL
• phospho-p38 MAPK (Thr180/Tyr182) Capture Antibody Reagent, 1 mL
• phospho-p38 MAPK (Thr180/Tyr182) Detection Antibody Reagent, 1 mL
• phospho-JNK 1/2 (Thr183/Tyr185) Capture Antibody Reagent, 1 mL
• phospho-JNK 1/2 (Thr183/Tyr185) Detection Antibody Reagent, 1 mL
• InstantOne Assay Plate, 96 well (sealed)
• Cell Lysis Buffer (5X), 10 mL
• Enhancer Solution, 1 mL
• Wash Buffer (10X), 15 mL
• Detection Reagent (TMB Substrate), 12 mL
• Stop Solution, 12 mL
• Positive Control Cell Lysate lyophilized, 0.25 mL
• Plate seal, 1 ea
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Cell Lysates
Target Species Human, Mouse
For Use With (Application) ELISA
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias MAPKMAPK; ERK; ERK-2; ERK2; ERT1; MAPK2; P42MAPK; PRKM1; PRKM2; p38; p40; p41; p41mapk; p42-MAPK;ERK-1; ERK1; ERT2; HS44KDAP; HUMKER1A; P44ERK1; P44MAPK; PRKM3; p44-ERK1; p44-MAPK;JNK; JNK-46; JNK1; JNK1A2; JNK21B1/2; PRKM8; SAPK1; SAPK1c;CSBP; CSBP1; CSBP2; CSPB1;
Gene ID (Entrez) 1432, 5594, 5595
Gene Symbol MAPK1, MAPK14, MAPK3
Kit Contents Capture Antibody, Detection Antibody Reagent, InstantOneTM Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate,20 μL
Storage Requirements 2-8°C
Synonym ERK, p38, JNK
Target JNK1/JNK2/JNK3/ERK1/2/p38 MAPK alpha
Test Time 1 hr 20 min
Total Assay Time 1 hr 10 min
Show More Show Less