Learn More
MAPA™ Ultrane™ Grip & Proof 500 Nitrile Coated Gloves
Grip & Proof coating enhances grip in oily environments, in addition to reducing muscle strain and boosting productivity.
Supplier:  MAPA™ 34500011
 View more versions of this product
Catalog No. 19-249-820
Only null left
Explore available promotions
Description
Description
  • Grip & Proof coating enhances grip in oily environments
  • Coating reduces muscle strain and boosts productivity
  • Repels oil penetration and reduces dermatitis
  • Color adapted to dirty environments for a prolonged durability
  • Silicone-free to prevent traces or defects on sheet metal and glass prior to painting
  • Washable up to 3 cleaning cycles without degradation
  • DMF-free
Specifications
Specifications
Nitrile
ANSI A1
Specific
11
Knit
11 in.
Black
No Texture
Seamless Knit Gloves
4121A
Ultrane 500
28 cm
Product Suggestions
Product Suggestions
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'