Learn More
MAPA™ Ultrane™ Grip & Proof 500 Nitrile Coated Gloves
Grip & Proof coating enhances grip in oily environments, in addition to reducing muscle strain and boosting productivity.
Supplier:  MAPA™ 34500048
 View more versions of this product
Catalog No. 19-249-817
Only null left
Explore available promotions
Description
Description
  • Grip & Proof coating enhances grip in oily environments
  • Coating reduces muscle strain and boosts productivity
  • Repels oil penetration and reduces dermatitis
  • Color adapted to dirty environments for a prolonged durability
  • Silicone-free to prevent traces or defects on sheet metal and glass prior to painting
  • Washable up to 3 cleaning cycles without degradation
  • DMF-free
Specifications
Specifications
Nitrile
ANSI A1
Specific
8
Knit
9.8 in.
Black
No Texture
Seamless Knit Gloves
4121A
Ultrane 500
25 cm
Product Suggestions
Product Suggestions