missing translation for 'onlineSavingsMsg'
Learn More

MAP4K4/HGK Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501560556
Encompass
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501560556 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501560556 Supplier Bethyl Laboratories Inc Supplier No. A301502AT

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

M4K4, a member of the STE20 subfamily of Ser/Thr protein kinases, may play a role in the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predomit in the liver, skeletal muscle and placenta.

Specifications

Antigen MAP4K4/HGK
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene MAP4K4
Gene Accession No. O95819
Gene Alias Mitogen-activated protein kinase kinase kinase kinase 4; MAPK/ERK kinase kinase kinase 4; MEK kinase kinase 4; HPK/GCK-like kinase HGK; Nck interacting kinase; MEKKK 4; MAP4K4; NIK; KIAA0687
Gene Symbols MAP4K4
Host Species Rabbit
Immunogen Between 550 and 600
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9448
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less