Learn More
Invitrogen™ Human MAP4K1 (HPK1), GST Tag Recombinant Protein
Recombinant Protein
$922.00 - $13920.00
Specifications
| Accession Number | Q92918 |
|---|---|
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50mM tris HCl with 0.25mM DTT, 0.1mM EDTA, 10mM glutathione, 0.1mM PMSF, 25% glycerol, 150mM NaCl and no preservative; pH 7.5 |
| Gene ID (Entrez) | 11184 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PV6355
|
Invitrogen™
PV6355 |
10 μg |
Each for $922.00
|
|
|||||
PV6356
|
Invitrogen™
PV6356 |
100 μg |
Each for $3,205.00
|
|
|||||
PV6357
|
Invitrogen™
PV6357 |
1 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Recombinant human MAP4K1 (1-346) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Validated in LanthaScreen™ Kinase Binding Assay Protocols available for simple use and optimized performance Multiple pack sizes available for convenient use Related Product:LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant, Full Length The gene accession number for MAP4K1 (HPK1) is NP_009112.1.
MAP4K1 (also known as HPK1) or Hematopoietic progenitor kinase 1 is a hematopoietic cell-restricted member of the Ste20 serine/threonine kinase super family. MAP4K1 is a tissue-specific upstream activator of the MEKK/JNK/SAPK signaling pathway. MAP4K1 diminishes T cell receptor (TCR) signaling activity and T cell proliferation by phosphorylating the adaptor protein SLP-76. The c-Jun amino-terminal kinases (JNKs)/stress-activated protein kinases (SAPKs) play a crucial role in stress responses in mammalian cells. The mechanism underlying this pathway in the hematopoietic system is unclear, but it is a key in understanding the molecular basis of blood cell differentiation. We have cloned a novel protein kinase, termed hematopoietic progenitor kinase 1 (HPK1), that is expressed predominantly in hematopoietic cells, including early progenitor cells. HPK1 is related distantly to the p21(Cdc42/Rac1)-activated kinase (PAK) and yeast STE20 implicated in the mitogen-activated protein kinase (MAPK) cascade. Expression of HPK1 activates JNK1 specifically, and it elevates strongly AP-1-mediated transcriptional activity in vivo. HPK1 binds and phosphorylates MEKK1 directly, whereas JNK1 activation by HPK1 is inhibited by a dominant-negative MEKK1 or MKK4/SEK mutant. Interestingly, unlike PAK65, HPK1 does not contain the small GTPase Rac1/Cdc42-binding domain and does not bind to either Rac1 or Cdc42, suggesting that HPK1.Specifications
| Q92918 | |
| Kinase Assay | |
| 11184 | |
| Human MAP4K1 (HPK1), GST Tag | |
| Purified | |
| RUO | |
| HPK1 | |
| 1-346 | |
| Unconjugated | |
| Recombinant | |
| 1-346 | |
| Serine/Threonine Kinase | |
| Kinases & Inhibitors | |
| Liquid |
| See Label | |
| 50mM tris HCl with 0.25mM DTT, 0.1mM EDTA, 10mM glutathione, 0.1mM PMSF, 25% glycerol, 150mM NaCl and no preservative; pH 7.5 | |
| 65 kDa | |
| 7.5 | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| EC 2.7.11.1; Hematopoietic progenitor kinase; hematopoietic progenitor kinase 1; HPK; HPK1; MAP4K1; MAPK/ERK kinase kinase kinase 1; MEK kinase kinase 1; MEKKK 1; mHPK1; mitogen activated protein kinase kinase kinase kinase 1; mitogen-activated protein kinase kinase kinase kinase 1 | |
| MAP4K1 | |
| Protein | |
| Human | |
| GST-tag | |
| -80°C, Avoid Freeze/Thaw Cycles | |
| Baculovirus | |
| Recombinant, Full Length |