Learn More
Thermo Scientific™ Manual Crimpers and Decrimpers for Sealing and Unsealing Crimp Vials
Catalog No. p-7237210
Change view
Click to view available options
For Use With (Equipment):
11 mm Crimp Vials
13 mm Crimp Vials
20 mm Crimp Vials
8 mm Crimp Vials
Type:
Manual Crimper
Manual Decrimper
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
03452414 11 mm Crimp Vials Manual Crimper
03452415 13 mm Crimp Vials Manual Crimper
03452416 20 mm Crimp Vials Manual Crimper
03452417 8 mm Crimp Vials Manual Crimper
03452418 11 mm Crimp Vials Manual Decrimper
03452419 13 mm Crimp Vials Manual Decrimper
03452420 20 mm Crimp Vials Manual Decrimper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 03452414 Supplier Thermo Scientific™ Supplier No. 60180CR11
Only null left
Use Thermo Scientific™ Manual Vial Crimpers and Decrimpers to attach or remove crimp caps from vials, respectively.
Use Thermo Scientific™ Vial Crimpers and Decrimpers to manually attach or remove crimp caps from vials, respectively. These manual crimpers and decappers are available in 8 mm, 11 mm, 13 mm, and 20 mm sizes and allow you to easily seal or remove crimp caps reliably.
Specifications
Type Manual Crimper
For Use With (Equipment) 11 mm Crimp Vials
Material Stainless Steel/Aluminum
Product Line SureSTART Level 2