Learn More
Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers
Catalog No. p-6152727
Change view
Click to view available options
Size:
8 mm
13 mm
20 mm
28 mm
30 mm
32 mm
Type:
Manual Crimper
Manual Crimper (Flip-off)
Manual Decrimper
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Type
0337575C 13 mm Manual Crimper (Flip-off)
0337575D 20 mm Manual Crimper (Flip-off)
0337575A 28 mm Manual Crimper
0337575B 32 mm Manual Crimper
0640619AD 8 mm Manual Decrimper
0337575F 28 mm Manual Decrimper
0337552C 30 mm Manual Decrimper
03377125 32 mm Manual Decrimper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 0337575C Supplier Thermo Scientific™ Supplier No. C4013100F
Only null left
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
Lightweight, precision crimpers attach seals to chromatography or serum vials.
  • Factory set to propery crimp standard-thickness seals
  • Adjustable handle setting scree allows proper crimps with septa of varying thickness
Specifications
Description Manual Crimper, Flip-Off
No. per Pack 1 Ea.
Type Manual Crimper (Flip-off)
For Use With (Equipment) National Vial, Chromacol Vial, Repeatable Crimp, Remove Seal, Attach Seals to Chromatography, Serum Vial
Size 13 mm
For Use With (Application) Industrial Chromatography
Product Line National
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.