missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, Powder, Multi-Compendial, U.S.P., J.T. Baker™

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
PubChem CID 6251
ChEBI CHEBI:16899
SMILES OCC(O)C(O)C(O)C(O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
02003296
View Documents Promotion Details
Avantor J.T.Baker
255301
500 g Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
02003295
View Documents Promotion Details
Avantor J.T.Baker
255305
2.5 kg Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

69-65-8
182.17
FBPFZTCFMRRESA-UHFFFAOYNA-N
6251
OCC(O)C(O)C(O)C(O)CO
C6H14O6
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
CHEBI:16899

Specifications

69-65-8
MFCD00064287
d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
OCC(O)C(O)C(O)C(O)CO
182.17
CHEBI:16899
Multi-Compendial/USP
Mannitol
C6H14O6
1L = 1.52kg
FBPFZTCFMRRESA-UHFFFAOYNA-N
(2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
6251
182.17
Powder
Videos
SDS
Documents

Documents