missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Mannitol (D) USP
Description
Mannitol (D) USP can be a white, crystalline powder or free-flowing granules. Is odorless.
Alternative name(s): D-mannitol,
Specifications
Specifications
| Chemical Name or Material | Mannitol (D) USP |
| Packaging | Jar, 300mL, White, HDPE, Round, 63-400, TE Pano Neck |
| CAS | 69-65-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 182.17 |
| Quantity | 100 g |
| Percent Purity | 97 to 102 % |
| Grade | USP |