missing translation for 'onlineSavingsMsg'
Learn More

Mannitol, ACS, MilliporeSigma™

Catalog No. MMX02141
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX02141 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX02141 Supplier Technidata Supplier No. MX02141

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity grade D-mannitol meeting ACS specifications for laboratory use.

Specifications

CAS 69-65-8
Melting Point 164°C to 169°C
Color White
Density 400 to 500kg/m3
Boiling Point 290°C to 295°C
Ignition Point 410°C
Packaging Poly Pail
Molecular Formula C6H14O6
MDL Number MFCD00064287
Quantity 3 kg
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES OCC(O)C(O)C(O)C(O)CO
IUPAC Name hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
PubChem CID 6251
ChEBI CHEBI:16899
Formula Weight 182.17
Physical Form Powder
Chemical Name or Material Mannitol, For The Determination of Boric Acid
Show More Show Less
Recommended Storage Store without restrictions