Learn More
Watson-Marlow Manifold Tubing - Silicone
Double pump segment manifold tubing — use one, transfer to the other for double life
Supplier: Watson-Marlow 983 0076 000
Description
- For use with 400DM2 and 400DM3 pumpheads
- Can be used with any multichannel cartridges
- Adjacent cartridges may contain tubing of a different material or size
Chemical Identifiers
| Silicone | |
| 0.8 mm | |
| Pump Tubing |
| 0.03 in. | |
| 400DM2 and 400DM3 pump heads and multichannel cartridge |
Specifications
| 1/50 in. | |
| Silicone | |
| 0.8 mm | |
| 0.76mm | |
| Black/Black |
| 0.76 mm | |
| 0.03 in. | |
| 400DM2 and 400DM3 pump heads and multichannel cartridge | |
| 0.07 to 4.6 (four roller pump), 0.06 to 3.9 (eight roller pump)mL/min. | |
| Pump Tubing |