missing translation for 'onlineSavingsMsg'
Learn More

Manidipine dihydrochloride

Catalog No. AAJ60496MD
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60496MD 250 mg
AAJ60496MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60496MD Supplier Thermo Scientific Chemicals Supplier No. J60496MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Calcium channel blocker | CAS: 89226-75-5 | C35H40Cl2N4O6 | 683.627 g/mol

Calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 89226-75-5
Molecular Formula C35H40Cl2N4O6
Molecular Weight (g/mol) 683.627
MDL Number MFCD00896434
InChI Key JINNGBXKBDUGQT-UHFFFAOYSA-N
Synonym manidipine hydrochloride
PubChem CID 150762
IUPAC Name 5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl

Specifications

Color Yellow
Quantity 250 mg
UN Number UN2811
Merck Index 14,5743
Solubility Information Soluble in DMSO; Slightly soluble in water and ethanol /n
Formula Weight 683.63
Physical Form Crystalline Powder
Chemical Name or Material Manidipine dihydrochloride
Hazard Category H301
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P310-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only