missing translation for 'onlineSavingsMsg'
Learn More

Manidipine Dihydrochloride 98.0+%, TCI America™

Catalog No. M22251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22251G Supplier TCI America Supplier No. M22251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89226-75-5
Molecular Formula C35H40Cl2N4O6
Molecular Weight (g/mol) 683.627
MDL Number MFCD00896434
InChI Key JINNGBXKBDUGQT-UHFFFAOYSA-N
Synonym manidipine hydrochloride
PubChem CID 150762
IUPAC Name 5-O-[2-(4-benzhydrylpiperazin-1-yl)ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;dihydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl.Cl

Specifications

Melting Point 211°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 683.63
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Manidipine Dihydrochloride
RTECSNumber US7975300
TSCA No