missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganous Sulfate Solution, For Dissolved Oxygen, Certified, LabChem™
$94.14 - $385.46
Specifications
| Color | Clear, colorless to pink soution |
|---|---|
| Format | Liquid |
| Packaging | Poly Bottle |
| Storage Requirements | Room Temperature |
| Type | Manganous Sulfate Solution |
Specifications
| Clear, colorless to pink soution | |
| Poly Bottle | |
| Manganous Sulfate Solution |
| Liquid | |
| Room Temperature |