missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganous Sulfate Solution, For Dissolved Oxygen, Certified, LabChem™
Specifications
Specifications
| Color | Clear, colorless to pink soution |
| Format | Liquid |
| Packaging | Poly Bottle |
| Quantity | 500 mL |
| Storage Requirements | Room Temperature |
| Type | Manganous Sulfate Solution |