missing translation for 'onlineSavingsMsg'
Learn More

Manganous Sulfate Solution, 364 g/L, Ricca Chemical

Catalog No. 462016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
462016 500 mL Natural Poly Bottle
462032 1 L Natural Poly Bottle
46201 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 462016 Supplier Ricca Chemical Company Supplier No. 462016

APHA Standard Methods Reagent

Chemical Identifiers

Molecular Formula H2MnO5S
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Color Pink
CAS 10034-96-5, 7732-18-5
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
Molecular Weight (g/mol) 169.01
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 364g/L
Chemical Name or Material Manganous Sulfate Solution