missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganous Sulfate Monohydrate, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| CAS Max % | 100.0 |
| Physical Form | Fine Crystals or Crystalline Powder |
| Quantity | 500 g |
| Percent Purity | 98 to 101% |
| Grade | ACS |
| Packaging | White Poly Wide Mouth Bottle |
| Chemical Name or Material | Manganous Sulfate, Monohydrate |
Safety and Handling
| ShelfLife | 60 months |