missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Manganous Sulfate, Monohydrate, ACS Grade, 98.0 to 101.0%, LabChem™
Supplier: LabChem LC165551
Specifications
850°C | |
700°C | |
10034-96-5 | |
100 | |
500 g | |
MnSO4·H2O | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
manganese(2+) hydrate sulfate | |
177577 | |
169.01 | |
ACS | |
Poly Bottle |
Manganous Sulfate | |
Monohydrate | |
Pink | |
Powder | |
H2MnO5S | |
MFCD00149159 | |
Soluble in water | |
O.[Mn++].[O-]S([O-])(=O)=O | |
169.01 | |
CHEBI:86364 | |
98.0 to 101.0% | |
Passes Test | |
Sulfur compounds |
Chemical Identifiers
10034-96-5 | |
169.01 | |
ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
177577 | |
manganese(2+) hydrate sulfate |
H2MnO5S | |
MFCD00149159 | |
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate | |
CHEBI:86364 | |
O.[Mn++].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement
May cause damage to organs (central nervous system) through prolonged or repeated exposure (inhalation).
Toxic to aquatic life with long lasting effects.
GHS P Statement
Do not breathe dust.
Avoid release to the environment.
Get medical advice and attention if you feel unwell.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature