missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganous Chloride, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 254005
Specifications
| Manganous Chloride | |
| 13446-34-9 | |
| 2.5 kg | |
| MFCD00149792 | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] | |
| 197.90 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 2.01kg |
| 4-Hydrate | |
| Crystals | |
| Cl2H8MnO4 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| manganese(2+) tetrahydrate dichloride | |
| 197.9 | |
| Poly Bottle |
Chemical Identifiers
| 13446-34-9 | |
| 197.90 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] |
| Cl2H8MnO4 | |
| MFCD00149792 | |
| manganese(2+) tetrahydrate dichloride |