missing translation for 'onlineSavingsMsg'
Learn More

Manganous Chloride, 4-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. p-5390788
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003376 500 g Poly Bottle
02003375 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02003376 Supplier Avantor Supplier No. 254001
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Crystals
Quantity 500 g
Formula Weight 197.9
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.01kg
Chemical Name or Material Manganous Chloride, 4-Hydrate