missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate, Monohydrate, Reagent, ACS, 98-101%, Spectrum™ Chemical

Catalog No. p-7157709
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
125 g
2.5 kg
4 x 2.5 kg
50 kg
500 g
6 x 500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18604214 125 g Poly Bottle
18604215 500 g Poly Bottle
18604217 2.5 kg Poly Pail
18604216 6 x 500 g Poly Bottle
18604218 4 x 2.5 kg Poly Pail
18604219 12 kg Poly Pail
18604220 50 kg Fiber Drum
7 options

Catalog No. 18604214

Supplier: Spectrum Chemical Mfg Cor M1115125GM

Only null left

M1115, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Reagent, ACS , is an inorganic compound that like many metal sulfates can form a variety of hydrates such as heptahydrates, monohydrates, tetrahydrates, pentahydrates. When these salts are dissolved, they tend to give a light pink solution, a characteristic of manganese sulfate. As an ACS grade Reagent, Spectrum Chemical manufactured manganese sulfate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 101%
Grade ACS
Loss on Ignition 10.0-12%
Packaging Poly Bottle