missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate, Monohydrate, Powder, FCC, BP, 98-102%, Spectrum™ Chemical

Catalog No. 18604204
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604204 500 g Poly Bottle
18604205 2.5 kg Poly Pail
18604206 12 kg Poly Pail
18604207 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604204 Supplier Spectrum Chemical Mfg Cor Supplier No. MA164500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

MA164, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Powder, FCC, BP - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 102%
Grade Multi-Compendial
Packaging Poly Bottle