missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, (Crystalline),MP Biomedicals

$77.75 - $77.75

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19470280
SDS View Documents
MP Biomedicals Inc
0219470280
100 g
Each for $77.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10034-96-5
169.01
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+) hydrate sulfate
H2MnO5S
MFCD00149159
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O

Specifications

10034-96-5
100 g
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+) hydrate sulfate
177577
169
Crystalline
H2MnO5S
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
O.[Mn++].[O-]S([O-])(=O)=O
169.01
CHEBI:86364
Manganese Sulfate Monohydrate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents