missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-32435
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M113500 500 g Poly Bottle
M1133 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M113500 Supplier Thermo Fisher Scientific Supplier No. M113 500
Only null left

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850°C
Melting Point 700°C
Color Pale Pink
Physical Form Solid
Quantity 500 g
Assay Percent Range 98.0 to 101.0 %
Formula Weight 169.02
Percent Purity 98.0 to 101.0%
Grade Certified ACS
Identification Pass Test
Loss on Ignition 10 to 12%
Packaging Poly Bottle
Decomposition Information 850°C
Density 3.25g/cm³
Chemical Name or Material Manganese Sulfate Monohydrate
Show More Show Less