missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate), Reagents

Catalog No. p-200020936
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337907 100 g
01337906 12 kg
01337905 3 kg
01337904 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337907 Supplier Labchem Inc Supplier No. C1243000100C2
Only null left

Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym Manganous Sulfate Monohydrate
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850°C
Melting Point 700°C
Color Pink
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.95
Chemical Name or Material Manganese Sulfate Monohydrate, ACS Reagent (Manganous Sulfate)