missing translation for 'onlineSavingsMsg'
Learn More

Manganese standard solution, for AAS, 1 mg/ml Mn in 2 to 5% HNO3

Catalog No. p-220927
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196111000 100 mL
AC196115000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC196111000 Supplier Thermo Scientific Chemicals Supplier No. 196111000
Only null left

CAS: 7439-96-5 | Mn | 54.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy

Specifications

Chemical Name or Material Manganese standard solution
Name Note 1 mg/ml Mn in 2% HNO3
Packaging Plastic bottle
CAS 7439-96-5, 7697-37-2, 7732-18-5
Density 1.0200g/mL
Specific Gravity 1.02
Physical Form Liquid
Molecular Formula Mn
Concentration 997 to 1003 μg/mL of Mn (2+) in 2% HNO3 (20°C)
MDL Number MFCD00011111
Color Colorless
Melting Point -3.0°C
Boiling Point 101.0°C
Solubility Soluble in water
InChI Key PWHULOQIROXLJO-UHFFFAOYSA-N
SMILES [Mn]
IUPAC Name manganese
Molecular Weight (g/mol) 54.94
Quantity 100 mL
PubChem CID 23930
ChEBI CHEBI:35154
Formula Weight 54.94
Grade AAS
Show More Show Less

RUO – Research Use Only