Learn More
Manganese Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$253.06 - $454.74
Chemical Identifiers
| CAS | 7439-96-5 |
|---|---|
| Molecular Formula | Mn |
| Molecular Weight (g/mol) | 54.94 |
| MDL Number | MFCD00011111 |
| InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
| PubChem CID | 23930 |
| ChEBI | CHEBI:35154 |
| IUPAC Name | manganese |
| SMILES | [Mn] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLMN2-2Y
|
SPEX CertiPrep Group LLC
PLMN22Y |
125 mL | Triple-leached LDPE bottle |
Each for $253.06
|
|
||||
|
PLMN2-2X
|
SPEX CertiPrep Group LLC
PLMN22X |
500 mL | Triple-leached LDPE bottle |
Each for $454.74
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Mn | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7439-96-5 | |
| 54.94 | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| CHEBI:35154 | |
| [Mn] |
| Mn | |
| MFCD00011111 | |
| 23930 | |
| manganese |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Mn | |
| AA, ICP | |
| Certified Reference Material | |
| [Mn] | |
| manganese | |
| 125 mL | |
| CHEBI:35154 | |
| Manganese Standard |
| 7439-96-5,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00011111 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 54.94 | |
| 23930 | |
| Assurance |
Safety and Handling
Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only