missing translation for 'onlineSavingsMsg'
Learn More

Manganese Metal, Flakes, 99.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor M109225KG

 View more versions of this product

Catalog No. 18611500

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Manganese Metal
1
Poly Bottle
54.94
Ungraded
7439-96-5
2.5 kg
Flake
99.5%

Chemical Identifiers

7439-96-5
54.94
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.