missing translation for 'onlineSavingsMsg'
Learn More

Manganese Metal, Flakes, 99.5%, Spectrum™ Chemical

Catalog No. p-7165826
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611501 500 g Poly Bottle
18611500 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18611501 Supplier Spectrum Chemical Mfg Cor Supplier No. M1092500GM
Only null left

Chemical Identifiers

CAS 7439-96-5
Molecular Weight (g/mol) 54.94

Specifications

CAS Max % 1
Quantity 500 g
Packaging Poly Bottle
Form Flake
Percent Purity 99.5%
Grade Ungraded
Chemical Name or Material Manganese Metal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.