missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 99%

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

Supplier:  Thermo Scientific Chemicals A1761536

 View more versions of this product

Catalog No. AAA1761536


Only null left
Explore available promotions

Description

Description

Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Manganese(II) sulfate monohydrate
3 to 3.5
H2MnO5S
UN3077
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese(2+);sulfate;hydrate
177577
169.02 (151.00 Anhydrous)
Odorless
2.95
10034-96-5
500 g
MFCD00149159
14,5739
Soluble in water and alcohol. Insoluble in ether.
O.[Mn++].[O-]S([O-])(=O)=O
169.01
CHEBI:86364
99%
Hygroscopic

Chemical Identifiers

10034-96-5
169.01
ISPYRSDWRDQNSW-UHFFFAOYSA-L
177577
manganese(2+);sulfate;hydrate
H2MnO5S
MFCD00149159
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
CHEBI:86364
O.[Mn++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

P260-P280-P305+P351+P338-P310-P314-P501c

H318-H373

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 232-089-9

RTECSNumber : OP0893500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only