missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 99+%, extra pure

Catalog No. p-3734914
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205905000 500 g Plastic bottle
AC205900010 1 kg Plastic bottle
AC205900050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC205905000 Supplier Thermo Scientific Chemicals Supplier No. 205905000
Only null left

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
IUPAC Name manganese(2+);sulfate;hydrate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

Boiling Point 850.0°C
Melting Point 700.0°C
Color Pink to White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnSO4·H2O
Merck Index 15, 5804
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 169.02
Percent Purity ≥99%
Grade Extra Pure
Loss on Ignition 10.0% to 12.0% (500°C)
Packaging Plastic bottle
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

RUO – Research Use Only