missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) sulfate monohydrate, 97%

Catalog No. p-7081720
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1180936 500 g
AAA118090E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1180936 Supplier Thermo Scientific Chemicals Supplier No. A1180936
Only null left

CAS: 10034-96-5 | H2MnO5S | 169.01 g/mol

Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

Solubility
Soluble in water. 5-10 g/100 mL at 21°C

Notes
Loses all water at 400-450°

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
Synonym manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
PubChem CID 177577
ChEBI CHEBI:86364
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

pH 3 to 3.5
Quantity 500 g
UN Number UN3077
Merck Index 14,5739
Solubility Information Soluble in water. 5-10g/100 mL at 21°C
IUPAC Name manganese(2+);sulfate;hydrate
Formula Weight 169.02 (151.00 Anhydrous)
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 2.95
Chemical Name or Material Manganese (II) sulfate monohydrate
Show More Show Less
Hazard Category H318-H373
Hazard Statement GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P280-P305+P351+P338-P310-P314-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 232-089-9
RTECSNumber OP0893500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only