Learn More
Manganese(II) fluoride, 99%
CAS: 7782-64-1 | F2Mn | 92.935 g/mol
Supplier: Thermo Scientific Chemicals 01152718
Description
Manganese(II) fluoride is used in the manufacture of special kinds of glass and lasers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Manganese(II) fluoride | |
| 7782-64-1 | |
| Crystalline Powder | |
| 99% | |
| MFCD00016222 | |
| Insoluble in water,alcohol. Soluble in dilute HF,conc. HCl or HNO3 | |
| F[Mn]F | |
| 92.935 | |
| 92.93 | |
| 3.98 |
| 856°C | |
| Red | |
| 50 g | |
| F2Mn | |
| manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis | |
| CTNMMTCXUUFYAP-UHFFFAOYSA-L | |
| difluoromanganese | |
| 24528 | |
| Hygroscopic |
Chemical Identifiers
| 7782-64-1 | |
| 92.935 | |
| CTNMMTCXUUFYAP-UHFFFAOYSA-L | |
| 24528 | |
| F[Mn]F |
| F2Mn | |
| MFCD00016222 | |
| manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis | |
| difluoromanganese |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : DOT Class: 6.1, Packaging Group: III
EINECSNumber : 231-960-0
RTECSNumber : OP0875000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only