missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) fluoride, 99%

Catalog No. p-4824809
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1152709 10 g
AA1152718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1152709 Supplier Thermo Scientific Chemicals Supplier No. 01152709
Only null left

CAS: 7782-64-1 | F2Mn | 92.935 g/mol

Manganese(II) fluoride is used in the manufacture of special kinds of glass and lasers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7782-64-1
Molecular Formula F2Mn
Molecular Weight (g/mol) 92.935
MDL Number MFCD00016222
InChI Key CTNMMTCXUUFYAP-UHFFFAOYSA-L
Synonym manganese ii fluoride, manganese fluoride mnf2, manganese fluorure french, manganese fluorure, mnf2, manganese fluoride di, manganese ii fluoride, anhydrous trace metals basis
PubChem CID 24528
IUPAC Name difluoromanganese
SMILES F[Mn]F

Specifications

Melting Point 856°C
Color Red
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99%
Solubility Information Insoluble in water,alcohol. Soluble in dilute HF,conc. HCl or HNO3
Formula Weight 92.93
Sensitivity Hygroscopic
Density 3.98
Chemical Name or Material Manganese(II) fluoride
Hazard Category H302+H312+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 231-960-0
RTECSNumber OP0875000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only