missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(Ii) Chloride Tetrahydrate, Honeywell Fluka™
Puriss. p.a., ACS Reagent,≥99%
Supplier: Honeywell Chemicals 314221KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Manganese(Ii) Chloride Tetrahydrate | |
| 1 kg | |
| MnCl2 · 4H2O | |
| NONH for all modes of transport | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] | |
| 197.90 | |
| ≥99% |
| 13446-34-9 | |
| Cl2H8MnO4 | |
| MFCD00149792 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| manganese(2+) tetrahydrate dichloride | |
| 197.91g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 13446-34-9 | |
| 197.90 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] |
| Cl2H8MnO4 | |
| MFCD00149792 | |
| manganese(2+) tetrahydrate dichloride |
Safety and Handling
P273
H302-H412
EINECSNumber : 231-869-6
RTECSNumber : OO9650000