missing translation for 'onlineSavingsMsg'
Learn More

Manganese(Ii) Chloride Tetrahydrate, Honeywell Fluka™

Catalog No. p-7072024
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002349 250 g
6002348 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6002349 Supplier Honeywell Chemicals Supplier No. 31422250G
Only null left

Puriss. p.a., ACS Reagent,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Quantity 250 g
Linear Formula MnCl2 · 4H2O
UN Number NONH for all modes of transport
Formula Weight 197.91g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a.
Chemical Name or Material Manganese(Ii) Chloride Tetrahydrate
Hazard Statement P273
Precautionary Statement H302-H412
EINECSNumber 231-869-6
RTECSNumber OO9650000