missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) chloride, 97%

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol

Supplier:  Thermo Scientific Chemicals 01186836

 View more versions of this product

Catalog No. AA1186836


Only null left
Explore available promotions

Description

Description

Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1190°C
650°C
Pink
500 g
Cl2Mn
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
GLFNIEUTAYBVOC-UHFFFAOYSA-L
manganese(2+) dichloride
10313134
125.84
Hygroscopic
Manganese(II) chloride
7773-01-5
Crystalline Flakes
97%
MFCD00011114
Soluble in water,pyridine and alcohol. Insoluble in ether.
[Cl-].[Cl-].[Mn++]
125.84
CHEBI:63041
Odorless
2.977

Chemical Identifiers

7773-01-5
125.84
GLFNIEUTAYBVOC-UHFFFAOYSA-L
10313134
[Cl-].[Cl-].[Mn++]
Cl2Mn
MFCD00011114
manganese chloride, manganese ii chloride, manganese 2+ ion dichloride, manganesechloride, acmc-20akkt, manganous chloride,anhydrous, ksc171o6h, manganous chloride, anhydr, manganese ii chloride anhydrous crystalline
CHEBI:63041
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c

H302-H318-H373

DOTInformation : DOT Class: 9; Packaging Group: III

EINECSNumber : 231-869-6

RTECSNumber : OO9625000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only