Learn More
Manganese(II) chloride, 97%
Description
Manganese(II) chloride, plays an important role in the production of dry cell batteries. It is also used as a starting material to the methylcyclopentadienyl manganese tricarbonyl, which has antiknock property. Further, it is used in the formation of an active Manganese(0) species useful for radical cyclization. It is also used in 31P-NMR to determine the size and lamellarity of phospholipidvesicles reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1190°C |
| Melting Point | 650°C |
| Color | Pink |
| Physical Form | Crystalline Flakes |
| Quantity | 500 g |
| Assay Percent Range | 97% |
| Solubility Information | Soluble in water,pyridine and alcohol. Insoluble in ether. |
| IUPAC Name | manganese(2+) dichloride |
| Formula Weight | 125.84 |
| Odor | Odorless |
| Show More |
Safety and Handling
| Hazard Category | H302-H318-H373 |
| Precautionary Statement | P262-P264-P270-P280i-P301+P312-P305+P351+P338-P310-P314-P330-P501c |
| DOTInformation | DOT Class: 9; Packaging Group: III |
| EINECSNumber | 231-869-6 |
| RTECSNumber | OO9625000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only