missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) 2,4-pentanedionate

Catalog No. p-7024322
Change view
Click to view available options
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
AAA1876214 25 g
AAA1876222 100 g
AAA1876236 500 g
3 options

Catalog No. AAA1876214

Supplier: Thermo Scientific Chemicals A1876214

Only null left

CAS: 14024-58-9 | C10H14MnO4 | 253.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C to 262°C (decomposition)
Density 1.6
Quantity 25 g
Formula Weight 253.16
Chemical Name or Material Manganese(II) 2,4-pentanedionate
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 237-858-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only