accessibility menu, dialog, popup

UserName

OOOOMnH3CH3CCH3CH3

CAS RN 14024-58-9

IUPAC Name: manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: manganese(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 253.16
Molecular Formula: C10H14MnO4
InChi Key: ZQZQURFYFJBOCE-FDGPNNRMSA-L
SMILES: [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)