missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) 2,4-pentanedionate

Catalog No. AAA1876214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1876214 25 g
AAA1876222 100 g
AAA1876236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1876214 Supplier Thermo Scientific Chemicals Supplier No. A1876214

CAS: 14024-58-9 | C10H14MnO4 | 253.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14024-58-9
Molecular Formula C10H14MnO4
Molecular Weight (g/mol) 253.16
MDL Number MFCD00000022 MFCD09998212
InChI Key ZQZQURFYFJBOCE-FDGPNNRMSA-L
Synonym bis 4-hydroxypent-3-en-2-one dihydrate manganese
PubChem CID 54669727
IUPAC Name (Z)-4-hydroxypent-3-en-2-one;manganese;dihydrate
SMILES [Mn++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 260°C to 262°C (decomposition)
Density 1.6
Quantity 25 g
Formula Weight 253.16
Chemical Name or Material Manganese(II) 2,4-pentanedionate
Hazard Category H315-H319-H335-H351
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 237-858-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only