missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Chloride Tetrahydrate, ACS Reagent, Reagents
Manganese Chloride Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$367.89 - $894.50
Chemical Identifiers
| CAS | 13446-34-9 |
|---|---|
| Molecular Formula | Cl2H8MnO4 |
| Molecular Weight (g/mol) | 197.90 |
| MDL Number | MFCD00149792 |
| InChI Key | CNFDGXZLMLFIJV-UHFFFAOYSA-L |
| Synonym | Manganous Chloride Tetrahydrate |
| IUPAC Name | manganese(2+) tetrahydrate dichloride |
| SMILES | O.O.O.O.[Cl-].[Cl-].[Mn++] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-909
|
Labchem Inc
C1241500125D2 |
125 g |
Each for $367.89
|
|
|||||
|
01-337-908
|
Labchem Inc
C1241500500C2 |
500 g |
Each for $894.50
|
|
|||||
Chemical Identifiers
| 13446-34-9 | |
| 197.90 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| manganese(2+) tetrahydrate dichloride |
| Cl2H8MnO4 | |
| MFCD00149792 | |
| Manganous Chloride Tetrahydrate | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] |
Specifications
| 1190°C | |
| 13446-34-9 | |
| 125 g | |
| MFCD00149792 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| manganese(2+) tetrahydrate dichloride | |
| ACS | |
| 2.01 |
| Pink | |
| Solid | |
| Cl2H8MnO4 | |
| Manganous Chloride Tetrahydrate | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] | |
| 197.90 | |
| Glass Bottle | |
| Manganese Chloride Tetrahydrate, ACS Reagent |