missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate, ACS Reagent, Reagents

Catalog No. 01337909
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337909 125 g
01337908 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337909 Supplier Labchem Inc Supplier No. C1241500125D2
Only null left

Manganese Chloride Tetrahydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
Synonym Manganous Chloride Tetrahydrate
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Boiling Point 1190°C
Color Pink
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.01
Chemical Name or Material Manganese Chloride Tetrahydrate, ACS Reagent