missing translation for 'onlineSavingsMsg'
Learn More

Manganese AA Standard, 1000 ppm Mn in 3% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company AMN1KH500

 View more versions of this product

Catalog No. AMN1KH-500


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Manganese AA Standard
7439-96-5,7647-01-0,7732-18-5
94.0,3.88,0.09
1 mL = 1 mg Mn
Mn
Colorless
PWHULOQIROXLJO-UHFFFAOYSA-N
manganese
500 mL
CHEBI:35154
Natural Poly Bottle
1,000ppm Mn
98.0,4.04,0.1
Liquid
MFCD00011111
100°C
[Mn]
54.94
23930

Chemical Identifiers

7439-96-5
54.94
PWHULOQIROXLJO-UHFFFAOYSA-N
CHEBI:35154
[Mn]
Mn
MFCD00011111
23930
manganese
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions