missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese AA Standard, 1000 ppm Mn in 3% HNO3, Ricca Chemical
$104.34 - $282.25
Chemical Identifiers
| CAS | 7439-96-5 |
|---|---|
| Molecular Formula | Mn |
| Molecular Weight (g/mol) | 54.94 |
| MDL Number | MFCD00011111 |
| InChI Key | PWHULOQIROXLJO-UHFFFAOYSA-N |
| PubChem CID | 23930 |
| ChEBI | CHEBI:35154 |
| IUPAC Name | manganese |
| SMILES | [Mn] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AMN1KN-100
|
Ricca Chemical Company
AMN1KN100 |
100 mL | Natural LDPE Bottle |
Each for $104.34
|
|
||||
|
AMN1KN-500
|
Ricca Chemical Company
AMN1KN500 |
500 mL | Natural Poly Bottle |
Each for $282.25
|
|
||||
Chemical Identifiers
| 7439-96-5 | |
| 54.94 | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| CHEBI:35154 | |
| [Mn] |
| Mn | |
| MFCD00011111 | |
| 23930 | |
| manganese |
Specifications
| Natural LDPE Bottle | |
| 1,000ppm Mn | |
| 98.0,4.04,0.1 | |
| Liquid | |
| MFCD00011111 | |
| < 2 | |
| PWHULOQIROXLJO-UHFFFAOYSA-N | |
| manganese | |
| 100 mL | |
| CHEBI:35154 |
| 7439-96-5,7647-01-0,7732-18-5 | |
| 94.0,3.88,0.09 | |
| 1 mL = 1 mg Mn | |
| Mn | |
| Colorless | |
| Approximately 0°C | |
| [Mn] | |
| 54.94 | |
| 23930 | |
| Manganese AA Standard |