missing translation for 'onlineSavingsMsg'
Learn More

Mancozeb, Thermo Scientific™

Catalog No. AC468850010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468850010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468850010 Supplier Thermo Scientific Chemicals Supplier No. 468850010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8018-01-7
Molecular Formula C8H12MnN4S8Zn
Molecular Weight (g/mol) 541.01
MDL Number 00078616
InChI Key CHNQZRKUZPNOOH-UHFFFAOYSA-J
IUPAC Name manganese(2+) zinc(2+) bis(({2-[(sulfanidylmethanethioyl)amino]ethyl}carbamothioyl)sulfanide)
SMILES [Mn++].[Zn++].[S-]C(=S)NCCNC([S-])=S.[S-]C(=S)NCCNC([S-])=S

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder
Color Yellow to Gray
Merck Index 13-5738
Quantity 1 g
Chemical Name or Material Mancozeb
Hazard Category Exclamation mark
Hazard Statement Health hazard
RTECSNumber ZB3200000
Recommended Storage Normal conditions